Learn More
Meso Scale Discovery UPLEX HUM PD1 epitope 2 ASYQP
SDP
Catalog No. 5000078519
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD1 epitope 2 in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L of sample and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance The kit contains reagents for one 96-well plate testing 40 samples in duplicate
Catalog No. 50-000-78519 Supplier Meso Scale Discovery Supplier No. K151V7K21
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD1 epitope 2 in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L of sample and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance The kit contains reagents for one 96-well plate testing 40 samples in duplicate