Learn More
Meso Scale Discovery UPLEX HUM PD1 epitope 2 ASYQP
SDP
Catalog No. 5000078977
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of PD1 epitope 2 biomarkers in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can monitor immune checkpoint inhibitor therapy by measuring free PD1 levels and anti-drug antibody responses It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance
Catalog No. 50-000-78977 Supplier Meso Scale Discovery Supplier No. K151V7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of PD1 epitope 2 biomarkers in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can monitor immune checkpoint inhibitor therapy by measuring free PD1 levels and anti-drug antibody responses It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance