Learn More
Meso Scale Discovery UPLEX HUM PDepitope 2 ASY384
SDP
Catalog No. 5000079854
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 384-Well Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD1 epitope 2 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides a 384-well plate formatted assay with high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit contains a PD1 epitope 2 calibrator detection antibody and streptavidin-gold It can facilitate biomarker research enable monitoring of immunotherapy responses and aid in drug development
Catalog No. 50-000-79854 Supplier Meso Scale Discovery Supplier No. K251V7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 384-Well Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD1 epitope 2 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides a 384-well plate formatted assay with high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit contains a PD1 epitope 2 calibrator detection antibody and streptavidin-gold It can facilitate biomarker research enable monitoring of immunotherapy responses and aid in drug development