Learn More
Meso Scale Discovery UPLEX HUM PDepitope ASY384
SDP
Catalog No. 5000080565
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 1 384-Well Assay Kit is designed to measure PD1 epitope 1 in human serum plasma and cell culture supernatants This electrochemiluminescence immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results in a 384-well format The kit includes a PD1 epitope 1 calibrator detection antibody and 384-well plates pre-coated with capture antibody for high throughput screening applications With a sample volume of 25 L and 3 5 hour total assay time this assay enables efficient quantification of PD1 epitope 1 levels
Catalog No. 50-000-80565 Supplier Meso Scale Discovery Supplier No. K251U7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD1 Epitope 1 384-Well Assay Kit is designed to measure PD1 epitope 1 in human serum plasma and cell culture supernatants This electrochemiluminescence immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results in a 384-well format The kit includes a PD1 epitope 1 calibrator detection antibody and 384-well plates pre-coated with capture antibody for high throughput screening applications With a sample volume of 25 L and 3 5 hour total assay time this assay enables efficient quantification of PD1 epitope 1 levels