Learn More
Meso Scale Discovery UPLEX HUM PDepitope ASY384well
SDP
Catalog No. 5000079403
Encompass
The U-PLEX Human PD1 Epitope 1 384-Well Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring PD1 epitope 1 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification with a broad dynamic range Requiring only 25 L of sample it detects as low as 19 pg/mL The kit includes a 384-well plate with anti-PD1 capture antibody detection antibody with ruthenium label and calibrators for standard curve generation offering a high-throughput method for research
Catalog No. 50-000-79403 Supplier Meso Scale Discovery Supplier No. K251U7K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human PD1 Epitope 1 384-Well Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring PD1 epitope 1 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification with a broad dynamic range Requiring only 25 L of sample it detects as low as 19 pg/mL The kit includes a 384-well plate with anti-PD1 capture antibody detection antibody with ruthenium label and calibrators for standard curve generation offering a high-throughput method for research