Learn More
Meso Scale Discovery UPLEX HUM PDL1 epitope 1 ASY
SDP
Catalog No. 5000079661
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 1) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD-L1 and other biomarkers from a single sample It uses electrochemiluminescence technology on patterned arrays for high sensitivity and broad dynamic range This assay kit includes a 96-well plate coated with capture antibodies for PD-L1 and other targets detection antibodies labeled with electrochemiluminescent tags calibrators and diluents optimized for human serum plasma and cell culture supernatant samples It provides a reproducible and sensitive way to measure PD-L1 expression alongside other immune checkpoint biomarkers
Catalog No. 50-000-79661 Supplier Meso Scale Discovery Supplier No. K151Z7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 1) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD-L1 and other biomarkers from a single sample It uses electrochemiluminescence technology on patterned arrays for high sensitivity and broad dynamic range This assay kit includes a 96-well plate coated with capture antibodies for PD-L1 and other targets detection antibodies labeled with electrochemiluminescent tags calibrators and diluents optimized for human serum plasma and cell culture supernatant samples It provides a reproducible and sensitive way to measure PD-L1 expression alongside other immune checkpoint biomarkers