Learn More
Meso Scale Discovery UPLEX HUM PDL1 epitope 1 ASY
SDP
Catalog No. 5000080484
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 1) Assay Kit is a multiplex immunoassay that measures PD-L1 in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls The assay has a typical sensitivity of 0 03 ng/mL and requires 50 L of sample
Catalog No. 50-000-80484 Supplier Meso Scale Discovery Supplier No. K151Z7K1
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 1) Assay Kit is a multiplex immunoassay that measures PD-L1 in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls The assay has a typical sensitivity of 0 03 ng/mL and requires 50 L of sample