Learn More
Meso Scale Discovery UPLEX HUM PDL1 epitope 1 ASYQP
SDP
Catalog No. 5000080481
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 1) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD-L1 and other biomarkers from a single sample It uses electrochemiluminescence technology on patterned arrays for high sensitivity and broad dynamic range This assay kit includes a 96-well plate coated with capture antibodies for PD-L1 and other targets along with detection antibodies and calibrators It provides a rapid reproducible method to measure PD-L1 expression levels in serum plasma cell culture supernatants and other sample types
Catalog No. 50-000-80481 Supplier Meso Scale Discovery Supplier No. K151Z7K21
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 1) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD-L1 and other biomarkers from a single sample It uses electrochemiluminescence technology on patterned arrays for high sensitivity and broad dynamic range This assay kit includes a 96-well plate coated with capture antibodies for PD-L1 and other targets along with detection antibodies and calibrators It provides a rapid reproducible method to measure PD-L1 expression levels in serum plasma cell culture supernatants and other sample types