Learn More
Meso Scale Discovery UPLEX HUM PDL1 epitope 2 ASYQP
SDP
Catalog No. 5000078862
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 2) Assay is a multiplex immunoassay kit for detecting and quantifying PD-L1 and other biomarkers from a single sample It uses electrochemiluminescence technology for high sensitivity and a broad dynamic range The kit includes reagents for measuring human PD-L1 and offers minimal hands-on time with rapid results using small sample volumes The U-PLEX platform allows customization of biomarker panels for specific research needs aiding in monitoring responses to cancer immunotherapies targeting the PD-1/PD-L1 pathway
Catalog No. 50-000-78862 Supplier Meso Scale Discovery Supplier No. K151AHSK22
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 2) Assay is a multiplex immunoassay kit for detecting and quantifying PD-L1 and other biomarkers from a single sample It uses electrochemiluminescence technology for high sensitivity and a broad dynamic range The kit includes reagents for measuring human PD-L1 and offers minimal hands-on time with rapid results using small sample volumes The U-PLEX platform allows customization of biomarker panels for specific research needs aiding in monitoring responses to cancer immunotherapies targeting the PD-1/PD-L1 pathway