Learn More
Meso Scale Discovery UPLEX HUM PDL2 ASYQP 25 PL
SDP
Catalog No. 5000079499
Encompass
The Meso Scale Discovery U-PLEX Human PD-L2 Assay is a multiplex immunoassay kit for measuring human PD-L2 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables rapid reproducible quantification of PD-L2 levels crucial for immunotherapy research
Catalog No. 50-000-79499 Supplier Meso Scale Discovery Supplier No. K151A8K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD-L2 Assay is a multiplex immunoassay kit for measuring human PD-L2 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables rapid reproducible quantification of PD-L2 levels crucial for immunotherapy research