Learn More
Meso Scale Discovery UPLEX HUM PDLepitope 2 ASY384
SDP
Catalog No. 5000078378
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 2) Assay 384-Well Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of Programmed Death Ligand 1 (PD-L1) in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of PD-L1 with an LLOQ of 19 5 pg/mL and requires as little as 25 L of sample This assay measures PD-L1 using antibodies targeting epitope 2 and can be used to monitor immune checkpoint inhibitor therapy response and PD-L1 expression
Catalog No. 50-000-78378 Supplier Meso Scale Discovery Supplier No. K251AHSK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 2) Assay 384-Well Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of Programmed Death Ligand 1 (PD-L1) in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of PD-L1 with an LLOQ of 19 5 pg/mL and requires as little as 25 L of sample This assay measures PD-L1 using antibodies targeting epitope 2 and can be used to monitor immune checkpoint inhibitor therapy response and PD-L1 expression