Learn More
Meso Scale Discovery UPLEX HUM PDLepitope 2 ASY384
SDP
Catalog No. 5000081136
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 2) Assay 384-Well Kit is a 384-well multiplex immunoassay that measures PD-L1 in human serum plasma and cell culture supernatant samples It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags and calibrators and controls needed to quantify PD-L1 levels It enables efficient and accurate measurement of PD-L1 for research use
Catalog No. 50-000-81136 Supplier Meso Scale Discovery Supplier No. K251AHSK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 2) Assay 384-Well Kit is a 384-well multiplex immunoassay that measures PD-L1 in human serum plasma and cell culture supernatant samples It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags and calibrators and controls needed to quantify PD-L1 levels It enables efficient and accurate measurement of PD-L1 for research use