Learn More
Meso Scale Discovery UPLEX HUM PDLepitope 2 ASYQP
SDP
Catalog No. 5000081616
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 2) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD-L1 and other biomarkers from a single sample It uses electrochemiluminescence technology on patterned arrays for high sensitivity and broad dynamic range This assay kit includes a plate coated with capture antibodies for PD-L1 and other targets along with detection antibodies and calibrators It allows quantification of PD-L1 levels in human serum plasma and cell culture supernatants for cancer immunotherapy research The assay has high specificity and sensitivity with minimal sample volume requirements
Catalog No. 50-000-81616 Supplier Meso Scale Discovery Supplier No. K151AHSK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 2) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD-L1 and other biomarkers from a single sample It uses electrochemiluminescence technology on patterned arrays for high sensitivity and broad dynamic range This assay kit includes a plate coated with capture antibodies for PD-L1 and other targets along with detection antibodies and calibrators It allows quantification of PD-L1 levels in human serum plasma and cell culture supernatants for cancer immunotherapy research The assay has high specificity and sensitivity with minimal sample volume requirements