Learn More
Meso Scale Discovery UPLEX HUM PDLepitope ASY384
SDP
Catalog No. 5000080562
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 Epitope 1 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of Programmed Death Ligand 1 (PD-L1) in human serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a rapid ultra-sensitive immunoassay with an extended dynamic range This product allows for the precise quantification of PD-L1 with a lower limit of detection of 49 5 pg/mL and an upper limit of 100 000 pg/mL The kit includes a PD-L1 calibrator and is for research use only
Catalog No. 50-000-80562 Supplier Meso Scale Discovery Supplier No. K251Z7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD-L1 Epitope 1 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of Programmed Death Ligand 1 (PD-L1) in human serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a rapid ultra-sensitive immunoassay with an extended dynamic range This product allows for the precise quantification of PD-L1 with a lower limit of detection of 49 5 pg/mL and an upper limit of 100 000 pg/mL The kit includes a PD-L1 calibrator and is for research use only