Learn More
Meso Scale Discovery UPLEX HUM PDLepitope ASY384
SDP
Catalog No. 5000080692
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 Epitope 1 384-Well Assay is an electrochemiluminescent immunoassay for measuring PD-L1 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible PD-L1 quantitation with a broad dynamic range Requiring only 25 L of sample it has a testing time of about 3 5 hours The kit includes a 384-well plate with PD-L1 capture antibody detection antibody read buffer sample diluent calibrators and controls ideal for high-throughput screening in research and drug development
Catalog No. 50-000-80692 Supplier Meso Scale Discovery Supplier No. K251Z7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD-L1 Epitope 1 384-Well Assay is an electrochemiluminescent immunoassay for measuring PD-L1 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible PD-L1 quantitation with a broad dynamic range Requiring only 25 L of sample it has a testing time of about 3 5 hours The kit includes a 384-well plate with PD-L1 capture antibody detection antibody read buffer sample diluent calibrators and controls ideal for high-throughput screening in research and drug development