Learn More
Meso Scale Discovery UPLEX HUM PDLepitope ASYQP
SDP
Catalog No. 5000078780
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 1) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD-L1 and other biomarkers from a single sample It uses electrochemiluminescence technology on patterned arrays for high sensitivity and broad dynamic range This assay kit includes a 96-well plate coated with capture antibodies for PD-L1 and other targets detection antibodies labeled with electrochemiluminescent tags calibrators controls 10X read buffer sample diluent wash buffer and plate seals It allows sensitive reproducible and quantitative measurement of PD-L1 levels in human serum plasma and other biological samples
Catalog No. 50-000-78780 Supplier Meso Scale Discovery Supplier No. K151Z7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 1) Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD-L1 and other biomarkers from a single sample It uses electrochemiluminescence technology on patterned arrays for high sensitivity and broad dynamic range This assay kit includes a 96-well plate coated with capture antibodies for PD-L1 and other targets detection antibodies labeled with electrochemiluminescent tags calibrators controls 10X read buffer sample diluent wash buffer and plate seals It allows sensitive reproducible and quantitative measurement of PD-L1 levels in human serum plasma and other biological samples