Learn More
Meso Scale Discovery UPLEX HUM Pentraxin 3 ASY
SDP
Catalog No. 5000079088
Encompass
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a multiplex immunoassay that measures Pentraxin 3 levels in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to enable the simultaneous detection of multiple protein biomarkers from a single sample The U-PLEX Pentraxin 3 Assay provides high sensitivity a broad dynamic range and requires low sample volumes This assay is designed for life science research use only and has not been tested for diagnostic purposes It is available as a singleplex assay in a 96-well plate format
Catalog No. 50-000-79088 Supplier Meso Scale Discovery Supplier No. K151AGZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a multiplex immunoassay that measures Pentraxin 3 levels in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to enable the simultaneous detection of multiple protein biomarkers from a single sample The U-PLEX Pentraxin 3 Assay provides high sensitivity a broad dynamic range and requires low sample volumes This assay is designed for life science research use only and has not been tested for diagnostic purposes It is available as a singleplex assay in a 96-well plate format