Learn More
Meso Scale Discovery UPLEX HUM Pentraxin 3 ASY384
SDP
Catalog No. 5000081117
Encompass
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of Pentraxin 3 in human serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a sensitive and robust assay with a broad dynamic range of 78 - 50 000 pg/mL The assay requires a low sample volume of 25 L and has high performance technical specifications including an intra-assay CV of 10% and inter-assay CV of 15% The kit includes a 10-plate kit with all necessary reagents and can quantify Pentraxin 3 in approximately 1 500 samples
Catalog No. 50-000-81117 Supplier Meso Scale Discovery Supplier No. K251AGZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of Pentraxin 3 in human serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a sensitive and robust assay with a broad dynamic range of 78 - 50 000 pg/mL The assay requires a low sample volume of 25 L and has high performance technical specifications including an intra-assay CV of 10% and inter-assay CV of 15% The kit includes a 10-plate kit with all necessary reagents and can quantify Pentraxin 3 in approximately 1 500 samples