Learn More
Meso Scale Discovery UPLEX HUM Pentraxin 3 ASYQP
SDP
Catalog No. 5000079089
Encompass
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a multiplex immunoassay kit that allows for the quantitative measurement of Pentraxin 3 in human serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires an MSD instrument and plates to run The kit includes a plate coated with capture antibodies a detection antibody conjugate calibrator and diluent The assay has a typical sensitivity of 0 04 ng/mL and an upper limit of quantification of 2500 ng/mL
Catalog No. 50-000-79089 Supplier Meso Scale Discovery Supplier No. K151AGZK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a multiplex immunoassay kit that allows for the quantitative measurement of Pentraxin 3 in human serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires an MSD instrument and plates to run The kit includes a plate coated with capture antibodies a detection antibody conjugate calibrator and diluent The assay has a typical sensitivity of 0 04 ng/mL and an upper limit of quantification of 2500 ng/mL