Learn More
Meso Scale Discovery UPLEX HUM Pentraxin ASY384
SDP
Catalog No. 5000081116
Encompass
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a 384-well immunoassay kit for the quantitative measurement of pentraxin 3 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 384-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator The assay has a typical lower limit of detection of 0 04 ng/mL and an upper limit of 10 000 ng/mL
Catalog No. 50-000-81116 Supplier Meso Scale Discovery Supplier No. K251AGZK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a 384-well immunoassay kit for the quantitative measurement of pentraxin 3 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 384-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator The assay has a typical lower limit of detection of 0 04 ng/mL and an upper limit of 10 000 ng/mL