Learn More
Meso Scale Discovery UPLEX HUM Perforin ASY 25 PL
SDP
Catalog No. 5000079086
Encompass
The Meso Scale Discovery U-PLEX Human Perforin Assay measures perforin levels in human serum plasma and cell culture supernatants using MSD s electrochemiluminescent technology on the MESO QuickPlex SQ 120 It allows simultaneous quantitative measurement of perforin crucial for immune response and cytolytic pathways with high sensitivity and a broad dynamic range The kit includes a 96-well plate capture and detection antibodies calibrators and controls Sample volume is 25 L and assay time is about 3 5 hours aiding in understanding immune function
Catalog No. 50-000-79086 Supplier Meso Scale Discovery Supplier No. K151AHGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Perforin Assay measures perforin levels in human serum plasma and cell culture supernatants using MSD s electrochemiluminescent technology on the MESO QuickPlex SQ 120 It allows simultaneous quantitative measurement of perforin crucial for immune response and cytolytic pathways with high sensitivity and a broad dynamic range The kit includes a 96-well plate capture and detection antibodies calibrators and controls Sample volume is 25 L and assay time is about 3 5 hours aiding in understanding immune function