Learn More
Meso Scale Discovery UPLEX HUM Perforin ASY384well
SDP
Catalog No. 5000078531
Encompass
The Meso Scale Discovery Human Perforin Singleplex Assay is an electrochemiluminescent immunoassay for the quantitative measurement of perforin in human serum plasma and cell culture supernatants It utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide a sensitive and reproducible method for measuring perforin levels This assay has a detection range of 3 05 - 10 000 pg/mL and requires 50 L sample size The total assay time is 3 5 hours This product can aid in understanding perforin s role in cytotoxic lymphocyte function and disease research
Catalog No. 50-000-78531 Supplier Meso Scale Discovery Supplier No. K251AHGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Human Perforin Singleplex Assay is an electrochemiluminescent immunoassay for the quantitative measurement of perforin in human serum plasma and cell culture supernatants It utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide a sensitive and reproducible method for measuring perforin levels This assay has a detection range of 3 05 - 10 000 pg/mL and requires 50 L sample size The total assay time is 3 5 hours This product can aid in understanding perforin s role in cytotoxic lymphocyte function and disease research