Learn More
Meso Scale Discovery UPLEX HUM Perforin ASY384well
SDP
Catalog No. 5000081005
Encompass
The Meso Scale Discovery U-PLEX Human Perforin Assay is a multiplex immunoassay that measures perforin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay allows for the simultaneous measurement of perforin along with other biomarkers It requires 50 L or less of sample and can perform up to 80 tests across two plates The plates are pre-coated with capture antibodies and do not require additional conjugation
Catalog No. 50-000-81005 Supplier Meso Scale Discovery Supplier No. K251AHGK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Perforin Assay is a multiplex immunoassay that measures perforin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay allows for the simultaneous measurement of perforin along with other biomarkers It requires 50 L or less of sample and can perform up to 80 tests across two plates The plates are pre-coated with capture antibodies and do not require additional conjugation