Learn More
Meso Scale Discovery UPLEX HUM Perforin ASYQP
SDP
Catalog No. 5000078864
Encompass
The Meso Scale Discovery U-PLEX Human Perforin Assay Kit is a multiplex immunoassay that measures human perforin in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use It can perform up to 80 tests in duplicate when used as directed
Catalog No. 50-000-78864 Supplier Meso Scale Discovery Supplier No. K151AHGK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Perforin Assay Kit is a multiplex immunoassay that measures human perforin in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use It can perform up to 80 tests in duplicate when used as directed