Learn More
Meso Scale Discovery UPLEX HUM PlGF ASY384well
SDP
Catalog No. 5000080276
Encompass
The Meso Scale Discovery U-PLEX Human PLGF 384-Well Assay Kit is a 384-well multiplex immunoassay that measures human placental growth factor (PLGF) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and assay diluent The assay takes approximately 3 5 hours to complete and requires a small sample volume of 25 L This quantitative immunoassay allows high-throughput analysis of PLGF levels
Catalog No. 50-000-80276 Supplier Meso Scale Discovery Supplier No. K251B8K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PLGF 384-Well Assay Kit is a 384-well multiplex immunoassay that measures human placental growth factor (PLGF) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and assay diluent The assay takes approximately 3 5 hours to complete and requires a small sample volume of 25 L This quantitative immunoassay allows high-throughput analysis of PLGF levels