Learn More
Meso Scale Discovery UPLEX HUM PlGF ASYQP 25 PL
SDP
Catalog No. 5000081606
Encompass
The Meso Scale Discovery U-PLEX Human PIGF Assay is a multiplex immunoassay kit for measuring human Placental Growth Factor (PIGF) in various biological samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls for generating a standard curve to quantify PIGF concentrations enabling rapid and efficient quantification of the angiogenesis biomarker PIGF
Catalog No. 50-000-81606 Supplier Meso Scale Discovery Supplier No. K151B8K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PIGF Assay is a multiplex immunoassay kit for measuring human Placental Growth Factor (PIGF) in various biological samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls for generating a standard curve to quantify PIGF concentrations enabling rapid and efficient quantification of the angiogenesis biomarker PIGF