Learn More
Meso Scale Discovery UPLEX HUM PP ASY384well
SDP
Catalog No. 5000080906
Encompass
The U-PLEX Human PP 384-Well Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures Properdin in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity and a wide dynamic range Requiring only 25 L per well the 384-well format supports high-throughput analysis The kit includes a microplate with capture antibodies detection antibodies buffers calibrators and controls for precise and accurate Properdin quantification
Catalog No. 50-000-80906 Supplier Meso Scale Discovery Supplier No. K2516AK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human PP 384-Well Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures Properdin in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity and a wide dynamic range Requiring only 25 L per well the 384-well format supports high-throughput analysis The kit includes a microplate with capture antibodies detection antibodies buffers calibrators and controls for precise and accurate Properdin quantification