Learn More
Meso Scale Discovery UPLEX HUM Proinsulin ASY384
SDP
Catalog No. 5000078371
Encompass
The Meso Scale Discovery U-PLEX Human Proinsulin 384-Well Assay Kit is a multiplex immunoassay that measures proinsulin levels in human serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay kit can help researchers investigate diabetes and metabolic disorders by enabling precise quantification of proinsulin levels
Catalog No. 50-000-78371 Supplier Meso Scale Discovery Supplier No. K2516MK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Proinsulin 384-Well Assay Kit is a multiplex immunoassay that measures proinsulin levels in human serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay kit can help researchers investigate diabetes and metabolic disorders by enabling precise quantification of proinsulin levels