Learn More
Meso Scale Discovery UPLEX HUM Proinsulin ASY384
SDP
Catalog No. 5000080918
Encompass
The Meso Scale Discovery U-PLEX Human Proinsulin 384-Well Assay Kit is a multiplex immunoassay that measures proinsulin levels in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of proinsulin The assay has a typical sensitivity of 0 04 pg/mL and an assay range from 0 08 - 1000 pg/mL The total assay time is 3 5 hours This assay kit offers a rapid ultra-sensitive method for quantifying proinsulin levels in research studies
Catalog No. 50-000-80918 Supplier Meso Scale Discovery Supplier No. K2516MK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Proinsulin 384-Well Assay Kit is a multiplex immunoassay that measures proinsulin levels in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of proinsulin The assay has a typical sensitivity of 0 04 pg/mL and an assay range from 0 08 - 1000 pg/mL The total assay time is 3 5 hours This assay kit offers a rapid ultra-sensitive method for quantifying proinsulin levels in research studies