Learn More
Meso Scale Discovery UPLEX HUM proMMP9 ASY384well
SDP
Catalog No. 5000080278
Encompass
The Meso Scale Discovery U-PLEX Human ProMMP-9 Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of proMMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can quantify proMMP-9 concentrations between 0 05-1500 ng/mL with an incubation time of 2 hours It includes a 384-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance
Catalog No. 50-000-80278 Supplier Meso Scale Discovery Supplier No. K251AHLK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ProMMP-9 Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of proMMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can quantify proMMP-9 concentrations between 0 05-1500 ng/mL with an incubation time of 2 hours It includes a 384-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance