Learn More
Meso Scale Discovery UPLEX HUM proMMP9 ASY384well
SDP
Catalog No. 5000081004
Encompass
The Meso Scale Discovery PromMP-9 Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for the quantitative measurement of pro-MMP-9 in serum plasma and cell culture supernatants The kit utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sub-picogram per milliliter detection sensitivity and a broad dynamic range The assay requires a 2 hour incubation time and has a lower limit of detection of 0 04 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and assay diluent
Catalog No. 50-000-81004 Supplier Meso Scale Discovery Supplier No. K251AHLK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery PromMP-9 Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for the quantitative measurement of pro-MMP-9 in serum plasma and cell culture supernatants The kit utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sub-picogram per milliliter detection sensitivity and a broad dynamic range The assay requires a 2 hour incubation time and has a lower limit of detection of 0 04 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and assay diluent