Learn More
Meso Scale Discovery UPLEX HUM proMMP9 ASYQP
SDP
Catalog No. 5000079523
Encompass
The Meso Scale Discovery U-PLEX Human ProMMP-9 Assay is a multiplex immunoassay for measuring ProMMP-9 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision Requiring 50 L or less sample volume it is compatible with the MESO QuickPlex SQ 120 instrument The U-PLEX platform measures biomarkers relevant to immunology oncology neurobiology and cardiovascular research providing rapid accurate results for advancing discovery in cancer metastasis arthritis and cardiovascular diseases
Catalog No. 50-000-79523 Supplier Meso Scale Discovery Supplier No. K151AHLK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ProMMP-9 Assay is a multiplex immunoassay for measuring ProMMP-9 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision Requiring 50 L or less sample volume it is compatible with the MESO QuickPlex SQ 120 instrument The U-PLEX platform measures biomarkers relevant to immunology oncology neurobiology and cardiovascular research providing rapid accurate results for advancing discovery in cancer metastasis arthritis and cardiovascular diseases