Learn More
Meso Scale Discovery UPLEX HUM PSelectin ASY
SDP
Catalog No. 5000079090
Encompass
The Meso Scale Discovery U-PLEX Human P-Selectin Assay is a multiplex immunoassay that measures P-selectin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The U-PLEX format allows simultaneous measurement of P-selectin with other biomarkers in human samples Overall this is a sensitive reproducible assay for quantifying P-selectin levels in various human sample types
Catalog No. 50-000-79090 Supplier Meso Scale Discovery Supplier No. K151AHNK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human P-Selectin Assay is a multiplex immunoassay that measures P-selectin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The U-PLEX format allows simultaneous measurement of P-selectin with other biomarkers in human samples Overall this is a sensitive reproducible assay for quantifying P-selectin levels in various human sample types