Learn More
Meso Scale Discovery UPLEX HUM PSelectin ASY384
SDP
Catalog No. 5000081120
Encompass
The Meso Scale Discovery V-PLEX Human P-Selectin Kit is an electrochemiluminescent immunoassay for measuring P-selectin in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for a sensitive assay detecting P-selectin at sub-picogram per mL with a range of 0 05-1000 ng/mL Intended for research use only it includes a 96-well plate with capture antibody detection antibody with ruthenium calibrators controls 10X diluent read buffer and plate seals for one 96-well plate
Catalog No. 50-000-81120 Supplier Meso Scale Discovery Supplier No. K251AHNK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human P-Selectin Kit is an electrochemiluminescent immunoassay for measuring P-selectin in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for a sensitive assay detecting P-selectin at sub-picogram per mL with a range of 0 05-1000 ng/mL Intended for research use only it includes a 96-well plate with capture antibody detection antibody with ruthenium calibrators controls 10X diluent read buffer and plate seals for one 96-well plate