Learn More
Meso Scale Discovery UPLEX HUM PSelectin ASY384well
SDP
Catalog No. 5000080917
Encompass
The Meso Scale Discovery U-PLEX Human P-Selectin Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of P-Selectin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires a Meso Scale Discovery SECTOR Imager and provides components for one 384-well microplate including antibody-coated plates calibrator detection antibody read buffer 10X wash buffer and plate seals The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 ng/mL
Catalog No. 50-000-80917 Supplier Meso Scale Discovery Supplier No. K251AHNK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human P-Selectin Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of P-Selectin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires a Meso Scale Discovery SECTOR Imager and provides components for one 384-well microplate including antibody-coated plates calibrator detection antibody read buffer 10X wash buffer and plate seals The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 ng/mL