Learn More
Meso Scale Discovery UPLEX HUM PYY TTL ASY384well
SDP
Catalog No. 5000078362
Encompass
The Meso Scale Discovery U-PLEX Human PYY Total 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of total Peptide YY (PYY) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This 384-well plate assay allows high-throughput analysis of total PYY levels It includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator diluent and lyophilized calibrator
Catalog No. 50-000-78362 Supplier Meso Scale Discovery Supplier No. K2516BK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PYY Total 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of total Peptide YY (PYY) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This 384-well plate assay allows high-throughput analysis of total PYY levels It includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator diluent and lyophilized calibrator