Learn More
Meso Scale Discovery UPLEX HUM RAGE soluble ASY
SDP
Catalog No. 5000081907
Encompass
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is a multiplex immunoassay kit for measuring RAGE in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It processes up to 80 samples in under 3 5 hours enhancing research workflow and conserving biological samples
Catalog No. 50-000-81907 Supplier Meso Scale Discovery Supplier No. K151AHAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is a multiplex immunoassay kit for measuring RAGE in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It processes up to 80 samples in under 3 5 hours enhancing research workflow and conserving biological samples