Learn More
Meso Scale Discovery UPLEX HUM RAGE soluble ASY
SDP
Supplier:  Meso Scale Discovery K151AHAK2
Encompass
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Receptor for Advanced Glycation Endproducts (RAGE) in human serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of soluble RAGE The assay has a detection range of 50-10 000 pg/mL and exhibits minimal cross-reactivity to related analytes Each kit provides sufficient reagents to perform up to 100 tests in a 96-well plate format This product can aid in studying inflammatory conditions where RAGE signaling plays a role
Catalog No. 50-000-78806