Learn More
Meso Scale Discovery UPLEX HUM RAGE soluble ASY
SDP
Catalog No. 5000079558
Encompass
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is an electrochemiluminescent immunoassay for measuring soluble receptor for advanced glycation endproducts (sRAGE) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and low sample volume Detection ranges from 1 15 pg/mL to 10 000 pg/mL Compatible with MSD instruments like SECTOR Imager and QuickPlex models it offers sensitive reproducible and rapid sRAGE measurement
Catalog No. 50-000-79558 Supplier Meso Scale Discovery Supplier No. K151AHAK1
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is an electrochemiluminescent immunoassay for measuring soluble receptor for advanced glycation endproducts (sRAGE) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and low sample volume Detection ranges from 1 15 pg/mL to 10 000 pg/mL Compatible with MSD instruments like SECTOR Imager and QuickPlex models it offers sensitive reproducible and rapid sRAGE measurement