Learn More
Meso Scale Discovery UPLEX HUM RAGE soluble ASY384
SDP
Catalog No. 5000080989
Encompass
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of Receptor for Advanced Glycation Endproducts (RAGE) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay allows for the simultaneous measurement of RAGE along with other biomarkers in the same sample With a testing time of 3 5 hours it enables rapid quantification of RAGE levels
Catalog No. 50-000-80989 Supplier Meso Scale Discovery Supplier No. K251AHAK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of Receptor for Advanced Glycation Endproducts (RAGE) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay allows for the simultaneous measurement of RAGE along with other biomarkers in the same sample With a testing time of 3 5 hours it enables rapid quantification of RAGE levels