Learn More
Meso Scale Discovery UPLEX HUM RAGE soluble ASY384
SDP
Catalog No. 5000081118
Encompass
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay 384-Well Kit is a multiplex immunoassay that measures soluble receptor for advanced glycation endproducts (sRAGE) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies Read Buffer and calibrator This assay allows for the quantitative measurement of sRAGE across a range of 78-50 000 pg/mL when using a 2-fold dilution series of calibrator
Catalog No. 50-000-81118 Supplier Meso Scale Discovery Supplier No. K251AHAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay 384-Well Kit is a multiplex immunoassay that measures soluble receptor for advanced glycation endproducts (sRAGE) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies Read Buffer and calibrator This assay allows for the quantitative measurement of sRAGE across a range of 78-50 000 pg/mL when using a 2-fold dilution series of calibrator