Learn More
Meso Scale Discovery UPLEX HUM RAGE soluble ASYQP
SDP
Catalog No. 5000081577
Encompass
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is an electrochemiluminescent immunoassay for measuring RAGE in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of RAGE involved in inflammation The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay enables high-throughput quantification of soluble RAGE levels
Catalog No. 50-000-81577 Supplier Meso Scale Discovery Supplier No. K151AHAK21
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is an electrochemiluminescent immunoassay for measuring RAGE in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of RAGE involved in inflammation The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay enables high-throughput quantification of soluble RAGE levels