Learn More
Meso Scale Discovery UPLEX HUM RAGE soluble ASYQP
SDP
Catalog No. 5000081578
Encompass
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is a multiplex immunoassay kit for measuring RAGE in human serum plasma and cell culture supernatants It uses electrochemiluminescent detection for high sensitivity up to 5-logs of dynamic range and low sample use The assay measures RAGE and other biomarkers simultaneously An MSD instrument like SECTOR Imager 2400 or 6000 is needed The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators and controls for standard curve generation and assay validation
Catalog No. 50-000-81578 Supplier Meso Scale Discovery Supplier No. K151AHAK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RAGE Soluble Assay is a multiplex immunoassay kit for measuring RAGE in human serum plasma and cell culture supernatants It uses electrochemiluminescent detection for high sensitivity up to 5-logs of dynamic range and low sample use The assay measures RAGE and other biomarkers simultaneously An MSD instrument like SECTOR Imager 2400 or 6000 is needed The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators and controls for standard curve generation and assay validation