Learn More
Meso Scale Discovery UPLEX HUM RANKL/TNFSF11 ASYQP
SDP
Catalog No. 5000080843
Encompass
The Meso Scale Discovery U-PLEX Human RANKL (TNFSF11) Assay Kit is a multiplex immunoassay that measures RANKL in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It can accurately quantify RANKL levels between 0 04-1000 pg/mL with minimal sample volume requirements This assay kit enables reliable reproducible analysis of RANKL levels in research studies
Catalog No. 50-000-80843 Supplier Meso Scale Discovery Supplier No. K151M7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RANKL (TNFSF11) Assay Kit is a multiplex immunoassay that measures RANKL in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It can accurately quantify RANKL levels between 0 04-1000 pg/mL with minimal sample volume requirements This assay kit enables reliable reproducible analysis of RANKL levels in research studies