Learn More
Meso Scale Discovery UPLEX HUM RANKL/TNFSF11 ASYQP
SDP
Catalog No. 5000082275
Encompass
The Meso Scale Discovery U-PLEX Human RANKL (TNFSF11) Assay Kit is a multiplex immunoassay that measures human RANKL in serum plasma cell culture supernatants and other biological samples This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls Results can be read on the MSD instrument platform and sample values are determined by comparing signals to a standard curve This assay allows for the quantitative measurement of human RANKL
Catalog No. 50-000-82275 Supplier Meso Scale Discovery Supplier No. K151M7K21
The Meso Scale Discovery U-PLEX Human RANKL (TNFSF11) Assay Kit is a multiplex immunoassay that measures human RANKL in serum plasma cell culture supernatants and other biological samples This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls Results can be read on the MSD instrument platform and sample values are determined by comparing signals to a standard curve This assay allows for the quantitative measurement of human RANKL