Learn More
Meso Scale Discovery UPLEX HUM RANKL/TNFSF1ASY384
SDP
Catalog No. 5000080568
Encompass
The Meso Scale Discovery U-PLEX Human RANKL (TNFSF11) 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous measurement of RANKL in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to quantify RANKL levels This assay can be run on MSD s SECTOR Imager 6000 for high throughput screening
Catalog No. 50-000-80568 Supplier Meso Scale Discovery Supplier No. K251M7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RANKL (TNFSF11) 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous measurement of RANKL in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to quantify RANKL levels This assay can be run on MSD s SECTOR Imager 6000 for high throughput screening