Learn More
Meso Scale Discovery UPLEX HUM RANKL/TNFSF1ASY384
SDP
Catalog No. 5000080733
Encompass
The Meso Scale Discovery U-PLEX Human RANKL (TNFSF11) Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of RANKL in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The kit includes a 384-well plate coated with an anti-RANKL capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80733 Supplier Meso Scale Discovery Supplier No. K251M7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RANKL (TNFSF11) Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of RANKL in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The kit includes a 384-well plate coated with an anti-RANKL capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL