Learn More
Meso Scale Discovery UPLEX HUM RANTES ASY 25 PL
SDP
Catalog No. 5000078961
Encompass
The Meso Scale Discovery U-PLEX Human RANTES Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of RANTES (CCL5) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology providing high sensitivity and a wide dynamic range The kit includes a plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer Diluent 100 and adhesive plate seals sufficient for one 96-well plate Recommended sample volume is 50 L The lower limit of detection is 0 64 pg/mL
Catalog No. 50-000-78961 Supplier Meso Scale Discovery Supplier No. K151A2K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RANTES Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of RANTES (CCL5) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology providing high sensitivity and a wide dynamic range The kit includes a plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer Diluent 100 and adhesive plate seals sufficient for one 96-well plate Recommended sample volume is 50 L The lower limit of detection is 0 64 pg/mL