Learn More
Meso Scale Discovery UPLEX HUM RANTES ASY384well
SDP
Catalog No. 5000081081
Encompass
The Meso Scale Discovery U-PLEX Human RANTES Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of RANTES in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires a Meso Scale Discovery SECTOR Imager instrument to read the plate The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for RANTES
Catalog No. 50-000-81081 Supplier Meso Scale Discovery Supplier No. K251A2K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RANTES Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of RANTES in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires a Meso Scale Discovery SECTOR Imager instrument to read the plate The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for RANTES