Learn More
Meso Scale Discovery UPLEX HUM RANTES ASY384well
SDP
Catalog No. 5000081105
Encompass
The Meso Scale Discovery U-PLEX Human RANTES Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of RANTES in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This multiplex assay allows simultaneous measurement of RANTES with other biomarkers in the same sample The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators and controls Sample incubation time is 2 hours
Catalog No. 50-000-81105 Supplier Meso Scale Discovery Supplier No. K251A2K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RANTES Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of RANTES in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This multiplex assay allows simultaneous measurement of RANTES with other biomarkers in the same sample The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators and controls Sample incubation time is 2 hours